Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172467.3 | NP_766055.1 | 2 | 436 | |
GenBank mRNA | AK045287.1 | - | 2 | 437 | |
AK051064.1 | - | 2 | 438 | ||
AK084475.1 | - | 2 | 368 | ||
BC138180.1 | - | 2 | 528 | 8861199 | |
BC138181.1 | - | 2 | 528 | 8861200 |