Gene Transcripts

Gene Symbol
Stxbp3
Entrez Gene ID
20912
Gene Name
syntaxin binding protein 3
Gene Aliases
Munc-18c, Stxbp3a, Sxtbp3, Unc18-3
Chromosome Location
Chr. 3: 108793180 - 108840502 on Build GRCm38
UniGene ID
Mm.316894
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011504.1 NP_035634.1 15 1312
GenBank mRNA AK138609.1 - 15 1344
AK138769.1 - 3 595
AK151466.1 - 15 1307
AK152261.1 - 15 1301
AK155133.1 - 15 1310
AK169799.1 - 14 1232
BC062901.1 - 15 1321 6810254
BC094055.1 - 15 1320
D30798.1 - 15 1312
U19521.1 - 15 1437

View Full Product Details