Gene Transcripts

Gene Symbol
Rsph4a
Entrez Gene ID
212892
Gene Name
radial spoke head 4 homolog A (Chlamydomonas)
Gene Aliases
A230081C05, Rshl3
Chromosome Location
Chr. 10: 33905111 - 33916021 on Build GRCm38
UniGene ID
Mm.131949
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162957.1 NP_001156429.1 2 931
GenBank mRNA AK038980.1 - 2 558
BC138000.1 - 2 931 8861019
BC138001.1 - 2 931 8861020

View Full Product Details