Gene Transcripts

Gene Symbol
Thbs2
Entrez Gene ID
21826
Gene Name
thrombospondin 2
Gene Aliases
TSP2, Thbs-2
Chromosome Location
Chr. 17: 14665500 - 14694262 on Build GRCm38
UniGene ID
Mm.26688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011581.3 NP_035711.2 20, 21 3759
GenBank mRNA AK132508.1 - 20, 21 3589
AK142552.1 - 20, 21 3731
AK165881.1 - 2, 3 376
BC031843.1 - 9, 10 1544
BC053702.1 - 20, 21 3561 30062369
L07803.1 - 20, 21 3587

View Full Product Details