Gene Transcripts

Gene Symbol
Lhfpl2
Entrez Gene ID
218454
Gene Name
lipoma HMGIC fusion partner-like 2
Gene Aliases
6030465B15, AI447312, AW050335, mKIAA0206, vgim
Chromosome Location
Chr. 13: 94057656 - 94195410 on Build GRCm38
UniGene ID
Mm.316553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172589.2 NP_766177.1 3 722
GenBank mRNA AK030103.1 - 2 880
AK031635.1 - 3 720
AK041685.1 - 1 379
AK052969.1 - 1 584
AK087506.1 - 3 723
BC040389.1 - 1 506 3484974

View Full Product Details