Gene Transcripts

Gene Symbol
Ucn
Entrez Gene ID
22226
Gene Name
urocortin
Gene Aliases
Mpv17, Ucn1
Chromosome Location
Chr. 5: 31137989 - 31138895 on Build GRCm38
UniGene ID
Mm.377116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021290.2 NP_067265.1 2 366
GenBank mRNA BC116948.1 - 2 300
BC116950.1 - 2 300 40126269
BC144891.1 - 2 295 9053439

View Full Product Details