Gene Transcripts

Gene Symbol
Timm8a2
Entrez Gene ID
223262
Gene Name
translocase of inner mitochondrial membrane 8A2
Gene Aliases
Chromosome Location
Chr. 14: 122028538 - 122038422 on Build GRCm38
UniGene ID
Mm.297637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037744.1 NP_001032833.1 1 58
GenBank mRNA BC099499.1 - 1 58

View Full Product Details