Gene Transcripts

Gene Symbol
Wnt10a
Entrez Gene ID
22409
Gene Name
wingless-type MMTV integration site family, member 10A
Gene Aliases
Chromosome Location
Chr. 1: 74792019 - 74804176 on Build GRCm38
UniGene ID
Mm.5130
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009518.2 NP_033544.1 3 544
GenBank mRNA BC014737.1 - 3 544
U61969.1 - 1054

View Full Product Details