Gene Transcripts

Gene Symbol
2410015M20Rik
Entrez Gene ID
224904
Gene Name
RIKEN cDNA 2410015M20 gene
Gene Aliases
QIL1, sr104
Chromosome Location
Chr. 17: 56607452 - 56609771 on Build GRCm38
UniGene ID
Mm.34002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153152.3 NP_694792.1 2 244
GenBank mRNA AK075942.1 - 2 175
AK075961.1 - 2 245
BC056164.1 - 2 150 5713062

View Full Product Details