Gene Transcripts

Gene Symbol
Fbxo2
Entrez Gene ID
230904
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, Fbs2, NFB42, Prpl4
Chromosome Location
Chr. 4: 148160226 - 148166417 on Build GRCm38
UniGene ID
Mm.262287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176848.1 NP_789818.1 3 485
GenBank mRNA AK140175.1 - 3 529
BC027053.1 - 2 438
BC046586.1 - 3 485 6487759

View Full Product Details