Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289727.1 | NP_001276656.1 | 4 | 478 | |
NM_001289728.1 | NP_001276657.1 | 4 | 478 | ||
NM_172930.4 | NP_766518.3 | 4 | 478 | ||
GenBank mRNA | AK041985.1 | - | 4 | 426 | |
AK045375.1 | - | 4 | 437 | ||
AK077178.1 | - | 4 | 413 | ||
AK078264.1 | - | 4 | 423 | ||
BC062956.1 | - | 4 | 394 | 30550174 |