Gene Transcripts
- Gene Symbol
- Mapkbp1
- Entrez Gene ID
- 26390
- Gene Name
- mitogen-activated protein kinase binding protein 1
- Gene Aliases
- 2810483F24Rik, AW123212, JNKBP-1, Jnkbp1, mKIAA0596
- Chromosome Location
- Chr. 2: 119972388 - 120027403 on Build GRCm38
- UniGene ID
- Mm.330501
- Species
- Mus musculus
- Gene Symbol
- Gm13999
- Entrez Gene ID
- 100036539
- Gene Name
- predicted gene 13999
- Gene Aliases
- OTTMUSG00000015282
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016792.1 |
- |
2 |
613 |
|
View Full Product Details