Gene Transcripts

Gene Symbol
Samd3
Entrez Gene ID
268288
Gene Name
sterile alpha motif domain containing 3
Gene Aliases
Gm623
Chromosome Location
Chr. 10: 26229618 - 26272916 on Build GRCm38
UniGene ID
Mm.213322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013766.2 NP_001013788.2 5 579
NM_001115154.1 NP_001108626.1 5 579
GenBank mRNA AK082201.1 - 5 580
BC150876.1 - 5 580 9087787

View Full Product Details