Gene Transcripts

Gene Symbol
Lppr1
Entrez Gene ID
272031
Gene Name
lipid phosphate phosphatase-related protein type 1
Gene Aliases
E130309F12Rik, PRG-3, mKIAA4247
Chromosome Location
Chr. 4: 49059462 - 49340261 on Build GRCm38
UniGene ID
Mm.299460
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178756.4 NP_848871.1 5 711
GenBank mRNA AK034251.1 - 5 806
AK043762.1 - 5 613
AK047614.1 - 5 641
AK087439.1 - 5 862
AK087514.1 - 5 712
AK160897.1 - 5 838
AY345342.1 - 5 711
BC096762.1 - 5 687 30094664

View Full Product Details