Gene Transcripts

Gene Symbol
Smok2a
Entrez Gene ID
27263
Gene Name
sperm motility kinase 2A
Gene Aliases
Smok2
Chromosome Location
Chr. 17: 13220998 - 13230264 on Build GRCm38
UniGene ID
Mm.371809
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013741.1 NP_038769.1 2 723
GenBank mRNA AJ245453.1 - 2 723

View Full Product Details