Gene Transcripts

Gene Symbol
Sycp2
Entrez Gene ID
320558
Gene Name
synaptonemal complex protein 2
Gene Aliases
3830402K23Rik, 4930518F03Rik
Chromosome Location
Chr. 2: 178345295 - 178407658 on Build GRCm38
UniGene ID
Mm.70781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177191.3 NP_796165.2 5 477
GenBank mRNA AK014411.1 - 5 505
DQ103262.1 - 5 477

View Full Product Details