Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289848.1 | NP_001276777.1 | 3 | 364 | |
NM_183405.2 | NP_899664.1 | 3 | 333 | ||
NM_183406.3 | NP_899665.1 | 2 | 244 | ||
GenBank mRNA | AK160152.1 | - | 3 | 263 | |
AY152400.1 | - | 3 | 333 | ||
BC048670.1 | - | 2 | 244 | 6773974 | |
BC107296.1 | - | 3 | 290 | 40053107 | |
BC107297.1 | - | 3 | 290 | 40053112 | |
BC125562.1 | - | 3 | 273 | 40130077 | |
BC125564.1 | - | 3 | 276 | 40130079 | |
BC145201.1 | - | 3 | 308 | 9053752 | |
CT010366.1 | - | 2 | 177 |