Gene Transcripts
- Gene Symbol
- Prickle4
- Entrez Gene ID
- 381104
- Gene Name
- prickle homolog 4 (Drosophila)
- Gene Aliases
- Gm939
- Chromosome Location
- Chr. 17: 47688474 - 47694958 on Build GRCm38
- UniGene ID
- Mm.392206
- Species
- Mus musculus
- Gene Symbol
- Tomm6os
- Entrez Gene ID
- 100415787
- Gene Name
- translocase of outer mitochondrial membrane 6, opposite strand
- Gene Aliases
- Gm14872, OTTMUSG00000018355, Prickle4os
- Chromosome Location
- Chr. 17: 47687610 - 47691092 on Build GRCm38
- UniGene ID
- Mm.392206
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK046990.1 |
- |
3 |
1299 |
|
View Full Product Details