Gene Transcripts

Gene Symbol
Serpinb3c
Entrez Gene ID
381286
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 3C
Gene Aliases
1110001H02Rik, 1110013A16Rik, Scca2, Serpinb4
Chromosome Location
Chr. 1: 107271201 - 107278371 on Build GRCm38
UniGene ID
Mm.337362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201363.2 NP_958751.2 6 607
GenBank mRNA AK003650.1 - 6 608
AY367775.1 - 5 537
BC132091.1 - 5 559 40130368

View Full Product Details