Gene Transcripts

Gene Symbol
Trim55
Entrez Gene ID
381485
Gene Name
tripartite motif-containing 55
Gene Aliases
D830041C10Rik, Murf2, Rnf29
Chromosome Location
Chr. 3: 19644328 - 19692348 on Build GRCm38
UniGene ID
Mm.35419
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081281.1 NP_001074750.1 1 199
GenBank mRNA AK052918.1 - 1 200

View Full Product Details