Gene Transcripts
- Gene Symbol
- Thap6
- Entrez Gene ID
- 381650
- Gene Name
- THAP domain containing 6
- Gene Aliases
- 4930534K21Rik, AI448650
- Chromosome Location
- Chr. 5: 91962882 - 91972137 on Build GRCm38
- UniGene ID
- Mm.159453
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015967.1 |
- |
3 |
948 |
|
BC089377.1 |
- |
2 |
260 |
|
View Full Product Details