Gene Transcripts

Gene Symbol
Cep44
Entrez Gene ID
382010
Gene Name
centrosomal protein 44
Gene Aliases
4933440G23Rik, mKIAA1712
Chromosome Location
Chr. 8: 56531516 - 56550777 on Build GRCm38
UniGene ID
Mm.425534
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009951.1 NP_001009951.1 9 1108
GenBank mRNA BC088983.1 - 9 1108

View Full Product Details