Gene Transcripts

Gene Symbol
Tmed8
Entrez Gene ID
382620
Gene Name
transmembrane emp24 domain containing 8
Gene Aliases
6430595O10Rik, AI447224, Gm1184, Mem1
Chromosome Location
Chr. 12: 87166242 - 87200229 on Build GRCm38
UniGene ID
Mm.374912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033475.3 NP_001028647.1 2 183
GenBank mRNA AK147378.1 - 2 184
BC150860.1 - 2 254 9087771

View Full Product Details