Gene Transcripts

Gene Symbol
Sval3
Entrez Gene ID
387564
Gene Name
seminal vesicle antigen-like 3
Gene Aliases
Chromosome Location
Chr. 6: 41968140 - 41973090 on Build GRCm38
UniGene ID
Mm.346825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003952.1 NP_001003952.1 2 145
GenBank mRNA AB098509.1 - 2 145

View Full Product Details