Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001495.2 | NP_001001495.2 | 2 | 272 | |
GenBank mRNA | AK033534.1 | - | 2 | 76 | |
BC104392.1 | - | 2 | 75 | ||
BC104393.1 | - | 2 | 75 | 40050409 | |
EF585494.1 | - | 2 | 254 | ||
EF585495.1 | - | 2 | 254 |