Gene Transcripts
- Gene Symbol
- Trgv2
- Entrez Gene ID
- 21636
- Gene Name
- T cell receptor gamma variable 2
- Gene Aliases
- Tcrg-V2, Vgamma2
- Chromosome Location
- Chr. 13: 19336573 - 19337026 on Build GRCm38
- UniGene ID
- Mm.211738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X00697.1 |
- |
|
958 |
|
View Full Product Details