Gene Transcripts
- Gene Symbol
- Xrra1
- Entrez Gene ID
- 446101
- Gene Name
- X-ray radiation resistance associated 1
- Gene Aliases
- AI449753
- Chromosome Location
- Chr. 7: 99859218 - 99917827 on Build GRCm38
- UniGene ID
- Mm.440757
- Species
- Mus musculus
- Gene Symbol
- Gm38405
- Entrez Gene ID
- 233561
- Gene Name
- predicted gene, 38405
- Gene Aliases
- A430054B03
- Chromosome Location
- Chr. 7: 99871365 - 99917373 on Build GRCm38
- UniGene ID
- Mm.234591
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040058.1 |
- |
|
435 |
|
View Full Product Details