Gene Transcripts
- Gene Symbol
- Cisd1
- Entrez Gene ID
- 52637
- Gene Name
- CDGSH iron sulfur domain 1
- Gene Aliases
- AU043990, AW743335, D10Ertd214e, Zcd1, mitoNEET
- Chromosome Location
- Chr. 10: 71330494 - 71344849 on Build GRCm38
- UniGene ID
- Mm.254114
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134007.4 |
NP_598768.1
|
1, 2 |
116 |
|
GenBank mRNA |
AK075907.1 |
- |
1, 2 |
117 |
|
AK131595.1 |
- |
1, 2 |
117 |
|
AK131605.1 |
- |
1, 2 |
95 |
|
AK131812.1 |
- |
1, 2 |
120 |
|
AK131995.1 |
- |
1, 2 |
127 |
|
AK167898.1 |
- |
1, 2 |
95 |
|
BC013522.1 |
- |
1, 2 |
105 |
4236502 |
BC019860.1 |
- |
1, 2 |
132 |
5123692 |
BC021952.1 |
- |
1, 2 |
153 |
5102315 |
View Full Product Details