Gene Transcripts

Gene Symbol
Arhgap40
Entrez Gene ID
545481
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, Gm14203, OTTMUSG00000015933
Chromosome Location
Chr. 2: 158512796 - 158550628 on Build GRCm38
UniGene ID
Mm.125371
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145015.1 NP_001138487.1 7, 8 1049

View Full Product Details