Gene Transcripts

Gene Symbol
Gramd2
Entrez Gene ID
546134
Gene Name
GRAM domain containing 2
Gene Aliases
BC064463
Chromosome Location
Chr. 9: 59679462 - 59718871 on Build GRCm38
UniGene ID
Mm.297580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033498.1 NP_001028670.1 1 234
GenBank mRNA AK041631.1 - 1 234
BC138948.1 - 1 51 8861970
BC138949.1 - 1 51 8861971

View Full Product Details