Gene Transcripts
- Gene Symbol
- Patz1
- Entrez Gene ID
- 56218
- Gene Name
- POZ (BTB) and AT hook containing zinc finger 1
- Gene Aliases
- 8430401L15Rik, AI447519, Mazr, Patz, Zfp278
- Chromosome Location
- Chr. 11: 3289131 - 3309083 on Build GRCm38
- UniGene ID
- Mm.275563
- Species
- Mus musculus
- Gene Symbol
- Gm12592
- Entrez Gene ID
- 791413
- Gene Name
- predicted gene 12592
- Gene Aliases
- OTTMUSG00000007639
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.437816
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029003.1 |
- |
2 |
419 |
|
View Full Product Details