Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190161.1 | NP_001177090.1 | 5 | 694 | |
NM_019976.3 | NP_064360.1 | 5 | 691 | ||
GenBank mRNA | AF156598.1 | - | 5 | 686 | |
AK017302.1 | - | 5 | 633 | ||
AK041835.1 | - | 5 | 641 | ||
AK167415.1 | - | 5 | 637 | ||
AK203468.1 | - | 2 | 94 | ||
BC008994.1 | - | 1 | 1494 | 3156760 | |
BC019165.1 | - | 5 | 649 | 3963608 |