Gene Transcripts

Gene Symbol
Stap1
Entrez Gene ID
56792
Gene Name
signal transducing adaptor family member 1
Gene Aliases
AI586015, Brdg1, STAP-1
Chromosome Location
Chr. 5: 86071585 - 86104784 on Build GRCm38
UniGene ID
Mm.131237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019992.3 NP_064376.1 2 162
GenBank mRNA AB036058.1 - 2 152
AK041474.1 - 2 163
AK042475.1 - 2 209
AK153661.1 - 2 241
AK162655.1 - 2 244
BC057898.1 - 2 216
BC145550.1 - 2 178 9054114
BC145828.1 - 2 177 40131131
BC145830.1 - 2 178 40131133

View Full Product Details