Gene Transcripts

Gene Symbol
Triml2
Entrez Gene ID
622117
Gene Name
tripartite motif family-like 2
Gene Aliases
EG622117, Tu4
Chromosome Location
Chr. 8: 43180506 - 43193884 on Build GRCm38
UniGene ID
Mm.390031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160412.1 NP_001153884.1 7 1238

View Full Product Details