Gene Transcripts

Gene Symbol
Ripply1
Entrez Gene ID
622473
Gene Name
ripply1 homolog (zebrafish)
Gene Aliases
Chromosome Location
Chr. X: 139779681 - 139782353 on Build GRCm38
UniGene ID
Mm.391312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037915.2 NP_001033004.1 4 407
GenBank mRNA AB212222.1 - 4 419

View Full Product Details