Gene Transcripts
- Gene Symbol
- Sdr39u1
- Entrez Gene ID
- 654795
- Gene Name
- short chain dehydrogenase/reductase family 39U, member 1
- Gene Aliases
- 2310014G06Rik
- Chromosome Location
- Chr. 14: 55897285 - 55900281 on Build GRCm38
- UniGene ID
- Mm.486218
- Species
- Mus musculus
- Gene Symbol
- Khnyn
- Entrez Gene ID
- 219094
- Gene Name
- KH and NYN domain containing
- Gene Aliases
- 6330555F21, 9130227C08Rik, AI605259, Kiaa0323, mKIAA0323
- Chromosome Location
- Chr. 14: 55884964 - 55898780 on Build GRCm38
- UniGene ID
- Mm.37592
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076856.1 |
- |
1 |
2324 |
|
AK122247.1 |
- |
8 |
4840 |
|
View Full Product Details