Gene Transcripts

Gene Symbol
Exosc3
Entrez Gene ID
66362
Gene Name
exosome component 3
Gene Aliases
2310005D06Rik, AI593501, Rrp40
Chromosome Location
Chr. 4: 45316629 - 45320603 on Build GRCm38
UniGene ID
Mm.275788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025513.3 NP_079789.1 3, 4 626
GenBank mRNA AK006281.1 - 3, 4 522
AK009165.1 - 1, 2 38
BC023669.1 - 3, 4 621 5012335
BC054085.1 - 3, 4 631 3371550

View Full Product Details