Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025598.2 | NP_079874.1 | 4 | 445 | |
GenBank mRNA | AK002602.1 | - | 3 | 565 | |
AK003282.1 | - | 4 | 446 | ||
AK012037.1 | - | 4 | 329 | ||
AK012327.1 | - | 4 | 447 | ||
AK012545.1 | - | 2 | 420 | ||
AK014211.1 | - | 3 | 399 | ||
AK154758.1 | - | 4 | 486 | ||
BC029225.1 | - | 3 | 381 | ||
BC058516.1 | - | 4 | 483 | 6824261 | |
BC091753.1 | - | 4 | 252 | 4218108 | |
BC100439.1 | - | 4 | 337 | 6531448 |