Gene Transcripts

Gene Symbol
Rwdd1
Entrez Gene ID
66521
Gene Name
RWD domain containing 1
Gene Aliases
0710001K08Rik, 2610002D06Rik, 2700069A07Rik, IH1
Chromosome Location
Chr. 10: 33996555 - 34019616 on Build GRCm38
UniGene ID
Mm.28528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025614.3 NP_079890.1 2 283
GenBank mRNA AF503942.1 - 2 175
AK003262.2 - 2 237
AK005278.1 - 2 195
AK009510.1 - 2 152
AK011277.1 - 2 283
AK012504.1 - 2 254
BC062136.1 - 2 165 6508909

View Full Product Details