Gene Transcripts

Gene Symbol
Diablo
Entrez Gene ID
66593
Gene Name
diablo homolog (Drosophila)
Gene Aliases
0610041G12Rik, 1700006L01Rik, AU040403, Smac
Chromosome Location
Chr. 5: 123511330 - 123524164 on Build GRCm38
UniGene ID
Mm.46716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023232.3 NP_075721.3 2 141
GenBank mRNA AF203914.1 - 2 107
AK002887.1 - 2 111
AK012760.1 - 2 139
AK077198.1 - 2 122
AK082903.1 - 2 111
AK133194.1 - 121
AK169573.1 - 2 111
BC024780.1 - 2 113 5365266
BC054109.1 - 2 135 30013833
BY080746.1 - 2 153

View Full Product Details