Gene Transcripts

Gene Symbol
Fignl2
Entrez Gene ID
668225
Gene Name
fidgetin-like 2
Gene Aliases
EG668225
Chromosome Location
Chr. 15: 101050192 - 101078584 on Build GRCm38
UniGene ID
Mm.385011
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001214911.2 NP_001201840.1 1 1211
GenBank mRNA BF464656.1 - 1 455
BU611164.1 - 1 18

View Full Product Details