Gene Transcripts

Gene Symbol
Ift27
Entrez Gene ID
67042
Gene Name
intraflagellar transport 27
Gene Aliases
2600013G09Rik, Rabl4
Chromosome Location
Chr. 15: 78159463 - 78174108 on Build GRCm38
UniGene ID
Mm.30191
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025931.2 NP_080207.1 5 574
GenBank mRNA AK011196.1 - 5 572
AK044278.1 - 2 1012
BC017514.1 - 5 574 4502942

View Full Product Details