Gene Transcripts

Gene Symbol
Uqcc2
Entrez Gene ID
67267
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 2
Gene Aliases
2900010M23Rik, Cbp6, M19, Mnf1
Chromosome Location
Chr. 17: 27122665 - 27133891 on Build GRCm38
UniGene ID
Mm.272389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026063.2 NP_080339.1 1, 2 142
GenBank mRNA AK013521.1 - 1, 2 146
AK019014.1 - 1, 2 143
BC030629.1 - 1, 2 145 1054665

View Full Product Details