Gene Transcripts

Gene Symbol
Cyp4f37
Entrez Gene ID
677156
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 37
Gene Aliases
Gm9705
Chromosome Location
Chr. 17: 32621319 - 32636184 on Build GRCm38
UniGene ID
Mm.484117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100187.1 NP_001093657.1 3 259
GenBank mRNA AK132492.1 - 3 259
BC151030.1 - 244 9087943

View Full Product Details