Gene Transcripts

Gene Symbol
Rbm42
Entrez Gene ID
68035
Gene Name
RNA binding motif protein 42
Gene Aliases
1700003D06Rik, 3100004P22Rik
Chromosome Location
Chr. 7: 30640995 - 30650228 on Build GRCm38
UniGene ID
Mm.401747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133693.2 NP_598454.2 10 1531
GenBank mRNA AK013929.1 - 10 1472
AK151000.1 - 10 1443
BC009148.1 - 10 1468 3600406
BC011286.1 - 10 1450 3485410
BC027372.1 - 10 1455 4952304
BC039917.1 - 10 1466
BC057928.1 - 10 1457 5289543
BY551742.1 - 3 331

View Full Product Details