Gene Transcripts

Gene Symbol
Ints1
Entrez Gene ID
68510
Gene Name
integrator complex subunit 1
Gene Aliases
1110015K06Rik
Chromosome Location
Chr. 5: 139751282 - 139775701 on Build GRCm38
UniGene ID
Mm.292942
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026748.2 NP_081024.3 10 1882
GenBank mRNA AB257854.1 - 10 1842
AK155838.1 - 10 1878
BC063266.1 - 10 1869 6409930

View Full Product Details