Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163476.1 | NP_001156948.1 | 2 | 222 | |
NM_027014.1 | NP_081290.1 | 2 | 222 | ||
GenBank mRNA | AK013116.1 | - | 2 | 215 | |
AK129081.1 | - | 1 | 13 | ||
BC027537.1 | - | 2 | 199 | 3466154 | |
BQ553304.1 | - | 2 | 222 |