Gene Transcripts
- Gene Symbol
- Wdfy1
- Entrez Gene ID
- 69368
- Gene Name
- WD repeat and FYVE domain containing 1
- Gene Aliases
- 1700013B03Rik, 1700120F24Rik, FENS-1, Jr1, WDF1, ZFYVE17, mKIAA1435
- Chromosome Location
- Chr. 1: 79702262 - 79776047 on Build GRCm38
- UniGene ID
- Mm.293273
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111279.1 |
NP_001104749.1
|
3, 4 |
348 |
|
NM_027057.3 |
NP_081333.1
|
3, 4 |
348 |
|
GenBank mRNA |
AK005937.1 |
- |
3, 4 |
312 |
|
AK007215.1 |
- |
3, 4 |
349 |
|
AK157204.1 |
- |
3, 4 |
307 |
|
AK159710.1 |
- |
3, 4 |
319 |
|
AK165554.1 |
- |
3, 4 |
320 |
|
AK220391.1 |
- |
3, 4 |
282 |
|
BC025226.1 |
- |
3, 4 |
262 |
5011404 |
BE197290.1 |
- |
2, 3 |
368 |
|
View Full Product Details