Gene Transcripts

Gene Symbol
Ctsll3
Entrez Gene ID
70202
Gene Name
cathepsin L-like 3
Gene Aliases
2310051M13Rik
Chromosome Location
Chr. 13: 60798250 - 60802844 on Build GRCm38
UniGene ID
Mm.158337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027344.3 NP_081620.2 6, 7 868
GenBank mRNA AK009935.1 - 1, 2 263
AK145382.1 - 6, 7 872
BC147423.1 - 5, 6 807 9056481
BC147424.1 - 5, 6 807 9056499

View Full Product Details