Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027994.1 | NP_082270.1 | 15 | 3917 | |
GenBank mRNA | AK009683.1 | - | 4 | 556 | |
AK078117.1 | - | 6 | 1144 | ||
AK129225.1 | - | 15 | 3878 | ||
BC027142.1 | - | 3 | 284 | ||
BC057457.1 | - | 8 | 2560 |